Tisk…

5-Chloro-2-pyridineboronic acid 95%

Dodavatel: Thermo Scientific
Varování

H52515.MD H52515.03
H52515.MDEA 2110 CZK
H52515.MD H52515.03
5-Chloro-2-pyridineboronic acid 95%
5-Chloro-2-pyridineboronic acid

Purity: 95% CAS No.: 652148-91-9 Molecular Formula: C5H5BClNO2 Formula Weight: 157.37 MDL No.: MFCD07368862

Vzorec: C₅H₅BClNO₂
MW: 157,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368862
CAS číslo: 652148-91-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR