4-Chloropicolinic acid ≥94%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H32301.03
H32301.09
H32301.03EA
1790
CZK
H32301.03
H32301.09
4-Chloropicolinic acid ≥94%
4-Chloropicolinic acid
Purity: 94%
CAS No.: 5470-22-4
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: ca 180° dec.
MDL No.: MFCD00191400
Vzorec:
C₆H₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00191400 CAS číslo: 5470-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...