5-Aminopicolinic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid
H27589.06
H27589.03
H27589.06EA
13040
CZK
H27589.06
H27589.03
5-Aminopicolinic acid 98%
5-Aminopicolinic acid
Purity: 98%
CAS No.: 24242-20-4
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Odour: Odourless
Melting Point: 217-219°
MDL No.: MFCD02684600
Vzorec:
C₆H₆N₂O₂ MW: 138.13 Bod tání: 217…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684600 CAS číslo: 24242-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...