2,6-Dichloro-3-pyridineboronic acid 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H27406.MD
H27406.03
H27406.MDEA
2780
CZK
H27406.MD
H27406.03
2,6-Dichloro-3-pyridineboronic acid 95%
2,6-Dichloro-3-pyridineboronic acid
Purity: 95%
CAS No.: 148493-34-9
Molecular Formula: C5H4BCl2NO2
Formula Weight: 191.81
MDL No.: MFCD03094989
Vzorec:
C₅H₄BCl₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03094989 CAS číslo: 148493-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...