5-Bromopicolinic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid
B25675.06
B25675.14
B25675.06EA
2550
CZK
B25675.06
B25675.14
5-Bromopicolinic acid ≥98%
5-Bromopicolinic acid
Purity: 98%
CAS No.: 30766-11-1
Molecular Formula: C6H4B2NO2
Formula Weight: 202.01
Melting Point: 173-175°
MDL No.: MFCD00234149
Vzorec:
C₆H₄BrNO₂ Bod tání: 173…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234149 CAS číslo: 30766-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...