N,N-Dimethylthioformamide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B24529.06
B24529.14
B24529.22
B24529.06EA
1250
CZK
B24529.06
B24529.14
B24529.22
N,N-Dimethylthioformamide ≥97%
N,N-Dimethylthioformamide
Purity: 97%
CAS No.: 758-16-7
Molecular Formula: C3H7NS
Formula Weight: 89.15
Boiling Point: 58-60°/1mm
MDL No.: MFCD00004943
Vzorec:
C₃H₇NS Bod varu: 58…60 °C (1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00004943 CAS číslo: 758-16-7 EINECS: 212-060-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...