Tisk…

Furfurylamine 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-(Aminomethyl)furan

B23975.22 B23975.36
B23975.22EA 1180 CZK
B23975.22 B23975.36
Furfurylamine 99%
Furfurylamine

Purity: 99% CAS No.: 617-89-0 Molecular Formula: C5H7NO Formula Weight: 97.12 Flash Point: 37°(99°F) Refractive Index: 1.4900 Odour: Amine-like pH: 11.6 Melting Point: -70° Boiling Point: 145-146° Beilstein Reference: 1614 UN No.: 2526 MDL No.: MFCD00003258

Vzorec: C₅H₇NO
MW: 97,12 g/mol
Bod varu: 145…146 °C
Bod tání: –70 °C
Hustota: 1,050
Bod vzplanutí: 37 °C (99 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003258
CAS číslo: 617-89-0
EINECS: 210-536-9
UN: 2526
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR