Tisk…

Tetraethylene glycol ≥99%

Dodavatel: Thermo Scientific

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol

B23880.0F B23880.AP
B23880.0FEA 3220 CZK
B23880.0F B23880.AP
Tetraethylene glycol ≥99%
Tetraethylene glycol

Purity: 99% CAS No.: 112-60-7 Molecular Formula: O(CH2CH2OCH2CH2OH)2 Formula Weight: 194.23 Flash Point: 193°(379°F) Refractive Index: 1.4590 Odour: Odourless pH: 8.5 Melting Point: -4° Boiling Point: 313-314° Beilstein Reference: 1634320 MDL No.: MFCD00002879

Vzorec: C₈H₁₈O₅
MW: 194,23 g/mol
Bod varu: 313…314 °C
Bod tání: –4 °C
Hustota: 1,125
Bod vzplanutí: 193 °C (379 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002879
CAS číslo: 112-60-7
EINECS: 203-989-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR