Diisopropyl phosphonate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite
B23800.14
B23800.22
B23800.14EA
1680
CZK
B23800.14
B23800.22
Diisopropyl phosphonate ≥98%
Diisopropyl phosphonate
Purity: 98%
CAS No.: 1809-20-7
Molecular Formula: C6H15O3P
Formula Weight: 166.16
Flash Point: 112°(234°F)
Refractive Index: 1.4075
Boiling Point: 72-75°/10mm
MDL No.: MFCD00117905
Vzorec:
C₆H₁₅O₃P Bod varu: 72…75 °C (10 mmHg) Hustota: 0,990 Bod vzplanutí: 112 °C (234 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00117905 CAS číslo: 1809-20-7 EINECS: 217-317-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...