Phenylphosphonic acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B22939.22EA
1710
CZK
B22939.22
Phenylphosphonic acid ≥98%
Phenylphosphonic acid
Vzorec:
C₆H₇O₃P MW: 158,09 g/mol Bod tání: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002136 CAS číslo: 1571-33-1 EINECS: 216-388-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...