Disodium benzene-1,3-disulphonate ≥94%
Dodavatel: Thermo Fisher Scientific
Synonyms:
1,3-Benzenedisulphonic acid disodium salt, Benzene-1,3-disulphonic acid disodium salt
B22811.14
B22811.22
B22811.36
B22811.14EA
731
CZK
B22811.14
B22811.22
B22811.36
Disodium benzene-1,3-disulphonate ≥94%
Disodium benzene-1,3-disulphonate
Purity: 94%
CAS No.: 831-59-4
Molecular Formula: C6H4Na2O6S2
Formula Weight: 282.20
MDL No.: MFCD00007492
Vzorec:
C₆H₄Na₂O₆S₂ MW: 282.21 Hustota: 1.124 Storage Temperature: Ambient |
MDL Number:
MFCD00007492 CAS číslo: 831-59-4 EINECS: 212-606-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...