Tisk…

Ethyl-5-bromonicotinate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B20788.06 B20788.14
B20788.06EA 3990 CZK
B20788.06 B20788.14
Ethyl-5-bromonicotinate ≥98%
Ethyl-5-bromonicotinate

Purity: 98% CAS No.: 20986-40-7 Molecular Formula: C8H8BrNO2 Formula Weight: 230.07 Melting Point: 39-42° Boiling Point: 86-92°/0.5mm MDL No.: MFCD00040366

Vzorec: C₈H₈BrNO₂
Bod varu: 86…92 °C (0,5 mmHg)
Bod tání: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00040366
CAS číslo: 20986-40-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR