4-Acetamidobenzaldehyde ≥98%
Dodavatel: Thermo Fisher Scientific
B20430.06
B20430.14
B20430.22
B20430.06EA
1500
CZK
B20430.06
B20430.14
B20430.22
4-Acetamidobenzaldehyde ≥98%
4-Acetamidobenzaldehyde
Purity: 98%
CAS No.: 122-85-0
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: 154-157°
MDL No.: MFCD00003380
Vzorec:
C₉H₉NO₂ Bod tání: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003380 CAS číslo: 122-85-0 EINECS: 204-579-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...