N,N-Dimethylglycine ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
A19670.06
A19670.22
A19670.14
A19670.06EA
2560
CZK
A19670.06
A19670.22
A19670.14
N,N-Dimethylglycine ≥98%
N,N-Dimethylglycine
Purity: 98+%
CAS No.: 1118-68-9
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Odour: Odourless
pH: 6-7
Melting Point: 179-183°
Merck Reference: 14,3244
MDL No.: MFCD00004283
Vzorec:
C₄H₉NO₂ MW: 103,12 g/mol Bod tání: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004283 CAS číslo: 1118-68-9 EINECS: 214-267-8 Merck Index: 14,3244 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...