Tisk…
N,N-Dimethylglycine hydrochloride ≥99%
Dodavatel: Thermo Scientific
Varování
Synonyms: N,N-Dimethylglycine HCl
A18892.06 A18892.14 A18892.22
A18892.06EA 1090 CZK
A18892.06 A18892.14 A18892.22
N,N-Dimethylglycine hydrochloride ≥99%
N,N-Dimethylglycine hydrochloride
Purity: 99% CAS No.: 2491-06-7 Molecular Formula: (CH3)2NCH2CO2H.HCl Formula Weight: 139.58 Melting Point: 189-194° Beilstein Reference: 3910578 Merck Reference: 14,3244 MDL No.: MFCD00012610
Vzorec: C₄H₁₀ClNO₂
MW: 139,58 g/mol
Bod tání: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00012610
CAS číslo: 2491-06-7
EINECS: 219-648-2
Merck Index: 14,3244