Tisk…

Acetylenedicarboxylic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A18736.06 A18736.14 A18736.22
A18736.06EA 1010 CZK
A18736.06 A18736.14 A18736.22
Acetylenedicarboxylic acid ≥97%
Acetylenedicarboxylic acid

Purity: 97% CAS No.: 142-45-0 Molecular Formula: HO2CC≡CCO2H Formula Weight: 114.06 Storage Temperature: 2°C to 8°C Flash Point: 131°(267°F) Melting Point: ca 182° dec. Beilstein Reference: 878357 UN No.: 2928 MDL No.: MFCD00004362

Vzorec: C₄H₂O₄
Bod vzplanutí: 131 °C (267 °F)
MDL Number: MFCD00004362
CAS číslo: 142-45-0
EINECS: 205-536-0
UN: 2928
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR