Tisk…

(±)-Ethyl mandelate ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl 2-hydroxy-2-phenylacetate

A17412.14 A17412.22
A17412.14EA 1370 CZK
A17412.14 A17412.22
(±)-Ethyl mandelate ≥97%
(±)-Ethyl mandelate

Purity: 97% CAS No.: 774-40-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Boiling Point: 253-255° MDL No.: MFCD00004494

Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Bod varu: 253…255 °C
Hustota: 1,115
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004494
CAS číslo: 774-40-3
EINECS: 212-263-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR