Tisk…

4-Methoxy-3-nitrobenzaldehyde ≥98%

Dodavatel: Thermo Scientific
Varování

A17081.06 A17081.14
A17081.06EA 1530 CZK
A17081.06 A17081.14
4-Methoxy-3-nitrobenzaldehyde ≥98%
4-Methoxy-3-nitrobenzaldehyde

Purity: 98% CAS No.: 31680-08-7 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 97-100° MDL No.: MFCD00126498

Vzorec: C₈H₇NO₄
MW: 181,15 g/mol
Bod tání: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00126498
CAS číslo: 31680-08-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR