Tisk…

Glyoxylic acid monohydrate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate

A16058.14 A16058.22
A16058.14EA 1180 CZK
A16058.14 A16058.22
Glyoxylic acid monohydrate ≥97%
Glyoxylic acid monohydrate

Purity: 97% CAS No.: 563-96-2 Molecular Formula: OHCCO2H.H2O Formula Weight: 92.06 (74.04anhy) Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 100° Beilstein Reference: 969535 Merck Reference: 14,4511 UN No.: 3261 MDL No.: MFCD00149497

Vzorec: C₂H₂O₃·1H₂O
Bod varu: 100 °C
Bod tání: 48…52 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006958
CAS číslo: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8,III
Merck Index: 14,4511

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR