Diphenyliodonium chloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A15149.09
A15149.18
A15149.30
A15149.09EA
3090
CZK
A15149.09
A15149.18
A15149.30
Diphenyliodonium chloride ≥98%
Diphenyliodonium chloride
Purity: 98+%
CAS No.: 1483-72-3
Molecular Formula: C12H10ClI
Formula Weight: 316.57
Melting Point: ca 230° dec.
Beilstein Reference: 3574977
Merck Reference: 14,7464
UN No.: 2811
MDL No.: MFCD00011909
Vzorec:
C₁₂H₁₀ClI MW: 316,57 g/mol Bod tání: ∼ 230 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011909 CAS číslo: 1483-72-3 EINECS: 216-049-8 UN: 2811 ADR: 6.1,III Merck Index: 14,7464 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...