Chloroacetamidine hydrochloride 96%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14835.06
A14835.14
A14835.06EA
3040
CZK
A14835.06
A14835.14
Chloroacetamidine hydrochloride 96%
Chloroacetamidine hydrochloride
Purity: 96%
CAS No.: 10300-69-3
Molecular Formula: C2H5ClN2.HCl
Formula Weight: 129.00
Storage Temperature: 2°C to 8°C
Melting Point: 97-103°
Beilstein Reference: 3678354
UN No.: 3261
MDL No.: MFCD00053013
Vzorec:
C₂H₆Cl₂N₂ Bod tání: 97…103 °C |
MDL Number:
MFCD00053013 CAS číslo: 10300-69-3 EINECS: 233-675-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...