Ethoxymethylenemalononitrile 97
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14412.14
A14412.22
A14412.36
A14412.14EA
1310
CZK
A14412.14
A14412.22
A14412.36
Ethoxymethylenemalononitrile 97
Ethoxymethylenemalononitrile
Purity: 97+%
CAS No.: 123-06-8
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Flash Point: 155°(311°F)
Melting Point: 62-68°
Boiling Point: 160°/12mm
Beilstein Reference: 1634241
UN No.: 3439
MDL No.: MFCD00001854
Vzorec:
C₆H₆N₂O MW: 122.13 Bod varu: 160 °C (12 mmHg) Bod tání: 62…68 °C Bod vzplanutí: 155 °C (311 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001854 CAS číslo: 123-06-8 EINECS: 204-597-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...