Cysteaminium chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A14377.14
A14377.22
A14377.36
A14377.14EA
1370
CZK
A14377.14
A14377.22
A14377.36
Cysteaminium chloride ≥98%
Cysteaminium chloride
Purity: 98+%
CAS No.: 156-57-0
Molecular Formula: HSCH2CH2NH2.HCl
Formula Weight: 113.61
Melting Point: 65-69°
Solubility: Soluble in water and alcohol
Beilstein Reference: 3590083
Merck Reference: 14,2779
MDL No.: MFCD00012904
Vzorec:
C₂H₇NS·HCl Bod tání: 65…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012904 CAS číslo: 156-57-0 EINECS: 205-858-1 Merck Index: 14,2779 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...