Tisk…

Diethyl acetamidomalonate ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Diethyl 2-acetamidomalonate

A14048.14 A14048.22 A14048.36
A14048.14EA 704 CZK
A14048.14 A14048.22 A14048.36
Diethyl acetamidomalonate ≥98%
Diethyl acetamidomalonate

Purity: 98+% CAS No.: 1068-90-2 Molecular Formula: CH3CONHCH(CO2CH2CH3)2 Formula Weight: 217.22 Melting Point: 96-99° Boiling Point: 185°/20mm MDL No.: MFCD00009146

Vzorec: C₉H₁₅NO₅
MW: 217.22
Bod varu: 185 °C (20 mmHg)
Bod tání: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00009146
CAS číslo: 1068-90-2
EINECS: 213-952-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR