3-Sulpholene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-sulpholene, Butadiene sulphone
A13887.0B
A13887.0I
A13887.22
A13887.36
A13887.0BEA
3880
CZK
A13887.0B
A13887.0I
A13887.22
A13887.36
3-Sulpholene ≥98%
3-Sulpholene
Purity: 98%
CAS No.: 77-79-2
Molecular Formula: C4H6O2S
Formula Weight: 118.15
Flash Point: 112°(233°F)
Melting Point: 63-67°
Beilstein Reference: 107004
Merck Reference: 14,8956
MDL No.: MFCD00005481
Vzorec:
C₄H₆O₂S Bod tání: 63…67 °C Hustota: 1,314 Bod vzplanutí: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005481 CAS číslo: 77-79-2 EINECS: 201-059-7 Merck Index: 14,8956 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...