Tisk…

N-(Hydroxymethyl)acetamide ≥98%

Dodavatel: Thermo Scientific

Synonyms: N-Hydroxymethylacetamide, N-Acetylmethanolamine, Acetamidomethanol

A13820.09 A13820.14
A13820.09EA 1870 CZK
A13820.09 A13820.14
N-(Hydroxymethyl)acetamide ≥98%
N-(Hydroxymethyl)acetamide

Purity: 98% CAS No.: 625-51-4 Molecular Formula: CH3CONHCH2OH Formula Weight: 89.09 Storage Temperature: 2°C to 8°C Melting Point: 47-52° Beilstein Reference: 506226 Merck Reference: 14,4242 MDL No.: MFCD00014417

Vzorec: C₃H₇NO₂
MW: 89,09 g/mol
Bod tání: 47…52 °C
Hustota: 1,14
MDL Number: MFCD00014417
CAS číslo: 625-51-4
EINECS: 210-897-2
Merck Index: 14,4242

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR