Tisk…

Phenylphosphonic dichloride ≥90%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13777.0B A13777.18 A13777.30
A13777.0BEA 13560 CZK
A13777.0B A13777.18 A13777.30
Phenylphosphonic dichloride ≥90%
Phenylphosphonic dichloride

Purity: 90+% CAS No.: 824-72-6 Molecular Formula: C6H5Cl2OP Formula Weight: 194.99 Flash Point: 204°(399°F) Refractive Index: 1.5600 Melting Point: 1-3° Boiling Point: 258° Beilstein Reference: 1072240 UN No.: 3265 MDL No.: MFCD00002070

Vzorec: C₆H₅Cl₂OP
Bod varu: 258 °C
Bod tání: 1…3 °C
Hustota: 1,379
Bod vzplanutí: 204 °C (399 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002070
CAS číslo: 824-72-6
EINECS: 212-534-3
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR