Diphenyl chlorophosphate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Diphenyl phosphorochloridate
A13546.0B
A13546.18
A13546.30
A13546.0BEA
11160
CZK
A13546.0B
A13546.18
A13546.30
Diphenyl chlorophosphate ≥97%
Diphenyl chlorophosphate
Purity: 97%
CAS No.: 2524-64-3
Molecular Formula: C12H10ClO3P
Formula Weight: 268.64
Flash Point: 113°(235°F)
Refractive Index: 1.5500
Boiling Point: 145-147°/1mm
Beilstein Reference: 654130
UN No.: 3265
MDL No.: MFCD00003030
Vzorec:
C₁₂H₁₀ClO₃P MW: 268.64 Bod varu: 145…147 °C (1 mmHg) Hustota: 1.296 Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003030 CAS číslo: 2524-64-3 EINECS: 219-759-6 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...