Cyanoacetamide ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Cyanoacetamide
A12482.0E
A12482.22
A12482.36
A12482.0EEA
13460
CZK
A12482.0E
A12482.22
A12482.36
Cyanoacetamide ≥99%
Cyanoacetamide
Purity: 99%
CAS No.: 107-91-5
Molecular Formula: NCCH2CONH2
Formula Weight: 84.08
Flash Point: 215°(419°F)
Melting Point: 118-122°
Beilstein Reference: 878221
Merck Reference: 14,2688
MDL No.: MFCD00008024
Vzorec:
C₃H₄N₂O MW: 84,08 g/mol Bod tání: 118…122 °C Bod vzplanutí: 215 °C (419 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008024 CAS číslo: 107-91-5 EINECS: 203-531-8 Merck Index: 14,2688 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...