Tisk…

1,8-Diazabicyclo[5.4.0]undec-7-ene ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12449.14 A12449.22 A12449.36
A12449.14EA 1450 CZK
A12449.14 A12449.22 A12449.36
1,8-Diazabicyclo[5.4.0]undec-7-ene ≥99%
1,8-Diazabicyclo[5.4.0]undec-7-ene

Purity: 99% CAS No.: 6674-22-2 Molecular Formula: C9H16N2 Formula Weight: 152.24 Flash Point: 116°(240°F) Refractive Index: 1.5220 Odour: Amine-like pH: 12.8 Melting Point: -70° Boiling Point: 261° Beilstein Reference: 508906 UN No.: 3267 MDL No.: MFCD00006930

Vzorec: C₉H₁₆N₂
Bod varu: 261 °C
Bod tání: –70 °C
Hustota: 1,020
Bod vzplanutí: 116 °C (240 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006930
CAS číslo: 6674-22-2
EINECS: 229-713-7
UN: 3267
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR