Bis(trimethylsilyl)acetylene ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A11960.09
A11960.18
A11960.30
A11960.09EA
1980
CZK
A11960.09
A11960.18
A11960.30
Bis(trimethylsilyl)acetylene ≥99%
Bis(trimethylsilyl)acetylene
Purity: 99%
CAS No.: 14630-40-1
Molecular Formula: C8H18Si2
Formula Weight: 170.40
Flash Point: 2°(35°F)
Refractive Index: 1.4270
Melting Point: 20-24°
Boiling Point: 134-136°
Beilstein Reference: 906870
UN No.: 1993
MDL No.: MFCD00008276
Vzorec:
C₈H₁₈Si₂ MW: 170.4 Bod varu: 134…136 °C Bod tání: 20…24 °C Hustota: 0.770 Bod vzplanutí: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008276 CAS číslo: 14630-40-1 EINECS: 238-671-9 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...