Tisk…

Diethyl acetylenedicarboxylate 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Diethyl but-2-yne-1,4-dioate

A11719.14 A11719.22 A11719.36
A11719.14EA 2560 CZK
A11719.14 A11719.22 A11719.36
Diethyl acetylenedicarboxylate 96%
Diethyl acetylenedicarboxylate

Purity: 96% CAS No.: 762-21-0 Molecular Formula: CH3CH2O2CC≡CCO2CH2CH3 Formula Weight: 170.16 Flash Point: 94°(201°F) Refractive Index: 1.4430 Melting Point: 1-2° Boiling Point: 108-110°/11mm Beilstein Reference: 743166 UN No.: 1760 MDL No.: MFCD00009186

Vzorec: C₈H₁₀O₄
MW: 170,16 g/mol
Bod varu: 108…110 °C (11 mmHg)
Bod tání: 1…2 °C
Hustota: 1,067
Bod vzplanutí: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009186
CAS číslo: 762-21-0
EINECS: 212-095-8
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR