Indium arsenide ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
22651.04
22651.09
22651.04EA
3250
CZK
22651.04
22651.09
Indium arsenide ≥99% (metals basis)
Indium arsenide
Purity: 99%
CAS No.: 1303-11-3
Molecular Formula: AsIn
Formula Weight: 189.74
Physical Form: 5mm & Down Pieces
Odour: Odourless
Merck Reference: 14,4949
UN No.: 1557
MDL No.: MFCD00016144
In semiconductor electronics
Vzorec:
AsIn Bod tání: 936 °C Hustota: 5,67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016144 CAS číslo: 1303-11-3 EINECS: 215-115-3 UN: 1557 ADR: 6.1,III Merck Index: 14,4949 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...