Tisk…

Dimethyl acetylenedicarboxylate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Dimethyl but-2-yne-1,4-dioate, Dimethyl but-2-ynedioate

A11437.14 A11437.22 A11437.36
A11437.14EA 1970 CZK
A11437.14 A11437.22 A11437.36
Dimethyl acetylenedicarboxylate ≥98%
Dimethyl acetylenedicarboxylate

Purity: 98% CAS No.: 762-42-5 Molecular Formula: C6H6O4 Formula Weight: 142.11 Flash Point: 86°(186°F) Refractive Index: 1.4480 Melting Point: -18° Boiling Point: 95-98°/19mm Beilstein Reference: 607063 UN No.: 1760 MDL No.: MFCD00008456

Vzorec: C₆H₆O₄
MW: 142,11 g/mol
Bod varu: 95…98 °C (19 mmHg)
Bod tání: –18 °C
Hustota: 1,156
Bod vzplanutí: 86 °C (186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008456
CAS číslo: 762-42-5
EINECS: 212-098-4
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR