Phenylarsonic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A10625.06
A10625.14
A10625.22
A10625.06EA
1060
CZK
A10625.06
A10625.14
A10625.22
Phenylarsonic acid ≥97%
Phenylarsonic acid
Purity: 97%
CAS No.: 98-05-5
Molecular Formula: C6H7AsO3
Formula Weight: 202.04
Melting Point: 158-162° dec.
Merck Reference: 14,1067
MDL No.: MFCD00002097
Vzorec:
C₆H₇AsO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00002097 CAS číslo: 98-05-5 EINECS: 202-631-9 UN: 3465 ADR: 6.1,III Merck Index: 14,1067 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...