Mercury(II) trifluoroacetate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
88132.06
88132.14
88132.06EA
1690
CZK
88132.06
88132.14
Mercury(II) trifluoroacetate ≥98%
Mercury(II) trifluoroacetate
Purity: 98+%
CAS No.: 13257-51-7
Molecular Formula: C4F6HgO4
Formula Weight: 426.62
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 167-169°
Beilstein Reference: 3633860
UN No.: 2025
MDL No.: MFCD00013201
Vzorec:
C₄F₆HgO₄ Bod tání: 167…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013201 CAS číslo: 13257-51-7 EINECS: 236-250-4 UN: 2025 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...