Dimethyl acetylenedicarboxylate ≥95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Dimethyl but-2-yne-1,4-dioate, Dimethyl but-2-ynedioate
45131.36
45131.22
45131.14
45131.36EA
24200
CZK
45131.36
45131.22
45131.14
Dimethyl acetylenedicarboxylate ≥95%
Dimethyl acetylenedicarboxylate
Purity: 95%
CAS No.: 762-42-5
Molecular Formula: C6H6O4
Formula Weight: 142.11
Flash Point: 86°(186°F)
Physical Form: Liquid
Density: 1.156
Refractive Index: 1.4470
Sensitivity: Light Sensitive
Appearance: Colourless to pale yellow
Melting Point: -18°
Boiling Point: 95-98°/19mm
Beilstein Reference: 607063
UN No.: 1760
MDL No.: MFCD00008456
Vzorec:
C₆H₆O₄ MW: 142,11 g/mol Bod varu: 95…98 °C (19 mmHg) Bod tání: –18 °C Hustota: 1,156 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008456 CAS číslo: 762-42-5 EINECS: 212-098-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...