Methacrolein ≥96% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with hydroquinone (HQ)
43124.AC
43124.AA
43124.ACEA
8270
CZK
43124.AC
43124.AA
Methacrolein ≥96% stabilizovaný
Methacrolein
Stabilized with hydroquinone
Purity: 96%
CAS No.: 78-85-3
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: -15°(5°F)
Physical Form: Liquid
Density: 0.847
Refractive Index: 1.4150
Sensitivity: Light Sensitive
Appearance: Pale yellow
Odour: Pungent
Melting Point: -81°
Boiling Point: 69-70°
UN No.: 2396
MDL No.: MFCD00006974
Vzorec:
C₄H₆O Bod varu: 69…70 °C Bod tání: –81 °C Hustota: 0,847 Bod vzplanutí: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006974 CAS číslo: 78-85-3 EINECS: 201-150-1 UN: 2396 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...