N,N-Dimethylsulfamoylchlorid 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Dimethylsulphamoyl chloride
408590250
408591000
408595000
ACRO408590250EA
2650
CZK
ACRO408590250
ACRO408591000
ACRO408595000
N,N-Dimethylsulfamoylchlorid 99%
N,N-Dimethylsulfamoylchlorid
Vzorec:
C₂H₆ClNO₂S MW: 143,59 g/mol Bod varu: 114 °C (75 mmHg) Bod tání: –13 °C Hustota: 1,337 g/cm³ Bod vzplanutí: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007425 CAS číslo: 13360-57-1 EINECS: 236-412-4 UN: 2927 ADR: 6.1,II |
Výsledky specifikačního testu
Appearance | Clear to hazy colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4530 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...