Tisk…

N,N-Dimethylallylamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N-Allyldimethylamine, 1-Dimethylamino-2-propene

407980250 407980050
ACRO407980250EA 5920 CZK
ACRO407980250 ACRO407980050
N,N-Dimethylallylamine 98%
N,N-Dimethylallylamine
Vzorec: C₅H₁₁N
MW: 85.15
Bod vzplanutí: 8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038234
CAS číslo: 2155-94-4
EINECS: 218-458-7
UN: 2733
ADR: 3 II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4000 to 1.4020 (20°C, 589 nm)
GC ≥97.5 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR