Diethyl chlorophosphate 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
407241000
407240250
407245000
ACRO407241000EA
3570
CZK
ACRO407241000
ACRO407240250
ACRO407245000
Diethyl chlorophosphate 95%
Diethyl chlorophosphate
Vzorec:
C₄H₁₀ClO₃P MW: 172.55 Bod varu: 81 °C (6 mmHg) Hustota: 1.19 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00009075 CAS číslo: 814-49-3 EINECS: 212-396-4 UN: 2927 ADR: 6.1 I |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4160 to 1.4180 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...