Tisk…

(R)-(+)-Hydroxypyrrolidine 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine

344570010 344570050
ACRO344570010EA 1890 CZK
ACRO344570010 ACRO344570050
(R)-(+)-Hydroxypyrrolidine 98%
(R)-(+)-Hydroxypyrrolidine
Vzorec: C₄H₉NO
MW: 87,12 g/mol
Bod varu: 108…110 °C (8 mmHg)
Bod vzplanutí: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00145220
CAS číslo: 2799-21-5

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to orange-brown Viscous liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4910 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +4.5° to +7.5° (20°C, 589 nm) (c=3.5, CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR