Tisk…

(S)-(+)-β-Aminophenethyl alcohol 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

307570010 307570050 307570250
ACRO307570010EA 1310 CZK
ACRO307570010 ACRO307570050 ACRO307570250
(S)-(+)-β-Aminophenethyl alcohol 98%
(S)-(+)-β-Aminophenethyl alcohol
Vzorec: C₈H₁₁NO
MW: 137.18
Bod tání: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS číslo: 20989-17-7
UN: 3263
ADR: 8 III

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 79 °C
HPLC ≥97.5 %
Specific optical rotation +24.5° to +26.5° (20 °C, 589 nm) (c=6, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR