Phosphinic acid (Hypophosphorous acid) 50% in water
Dodavatel: Thermo Scientific
|
Nebezpečí
|
201000010
201000050
201005001
ACRO201000010EA
7840
CZK
ACRO201000010
ACRO201000050
ACRO201005001
Phosphinic acid (Hypophosphorous acid) 50% in water
Phosphinic acid (Hypophosphorous acid)
Vzorec:
H₃O₂P MW: 66 g/mol Bod varu: 108 °C (1013hPa) Bod tání: –25 °C Hustota: 1,27 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD02183592 CAS číslo: 6303-21-5 EINECS: 228-601-5 UN: 3264 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless Solution |
Titration with NaOH | 48.0 to 52.0 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...