Ethylenediammonium dichloride 98+%
Dodavatel: Thermo Scientific
|
Varování
|
187651000
187655000
187650025
ACRO187651000EA
1290
CZK
ACRO187651000
ACRO187655000
ACRO187650025
Ethylenediammonium dichloride 98+%
Ethylenediammonium dichloride
Vzorec:
C₂H₁₀Cl₂N₂ MW: 133,02 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012524 CAS číslo: 333-18-6 EINECS: 206-369-6 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder, crystals or granules |
Infrared spectrum | Conforms |
Solubility | (1 % in water) Clear colorless |
pH | 3.9 to 5 (1 % in water) |
Titration with NaOH | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...