Iodosobenzene diacetate 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
176560250
176561000
176565000
ACRO176560250EA
2760
CZK
ACRO176560250
ACRO176561000
ACRO176565000
Iodosobenzene diacetate 98%
Iodosobenzene diacetate
Vzorec:
C₁₀H₁₁IO₄ Bod tání: 161…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008692 CAS číslo: 3240-34-4 EINECS: 221-808-1 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 159 to 164 °C |
Titration Iodometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...