Hydantoin 99%
Dodavatel: Thermo Scientific
Synonyms:
Imidazolidine-2,4-dione
168701000
168705000
ACRO168701000EA
1120
CZK
ACRO168701000
ACRO168705000
Hydantoin 99%
Hydantoin
Vzorec:
C₃H₄N₂O₂ MW: 100,08 g/mol Bod tání: 220…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005259 CAS číslo: 461-72-3 EINECS: 207-313-3 |
Výsledky specifikačního testu
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 220 to 223 °C |
Titration with NaOH | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...