Sodium benzenesulphinate 97%
Dodavatel: Thermo Fisher Scientific
154051000
154055000
ACRO154051000EA
2120
CZK
ACRO154051000
ACRO154055000
Sodium benzenesulphinate 97%
Sodium benzenesulphinate
Vzorec:
C₆H₅NaO₂S Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013135 CAS číslo: 873-55-2 EINECS: 212-842-8 |
Výsledky specifikačního testu
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | 96.0 to 104.0 % |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...