Diethyl phenylmalonate 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Diethyl 2-phenylmalonate
150260010
150261000
150262500
150260025
150260010.EA
5080
CZK
150260010.
150261000.
150262500.
150260025.
Diethyl phenylmalonate 98%
Diethyl phenylmalonate
Vzorec:
C₁₃H₁₆O₄ Bod varu: 170…172 °C (14 mmHg) Bod tání: 16…17 °C Hustota: 1,09 g/cm³ Bod vzplanutí: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009144 CAS číslo: 83-13-6 EINECS: 201-456-5 |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.4903 to 1.4923 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...