DL-α-Phenylglycine 99%
Dodavatel: Thermo Scientific
Synonyms:
Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine
130641000
130645000
ACRO130641000EA
1020
CZK
ACRO130641000
ACRO130645000
DL-α-Phenylglycine 99%
DL-α-Phenylglycine
Vzorec:
C₈H₉NO₂ MW: 151,16 g/mol Bod tání: 290 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00064402 CAS číslo: 2835-06-5 EINECS: 220-608-1 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Appearance of solution | (5% in 1N HCl) clear colorless to slightly yellow |
Titration with HClO4 | ≥98.5 % |
UV | at 200 nm A: ≤0.010 |
Gradient analysis | Passes test |
Absorbance | Passes test |
KMnO4-reducing substance | passes test |
Conductivity | 2.0 x 10-6 ohm-1 cm-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...